Iminodiacetic acid, 98+%
Iminodiacetic acid, 98+%
Iminodiacetic acid, 98+%
Iminodiacetic acid, 98+%
Thermo Scientific Chemicals
Iminodiacetic acid, 98+%
CAS: 142-73-4 | C4H7NO4 | 133.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number FSA204975000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIminodiacetic acid
Name Note98+%
CAS142-73-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: