1-Fluoro-4-iodobenzene, 99%
1-Fluoro-4-iodobenzene, 99%
1-Fluoro-4-iodobenzene, 99%
1-Fluoro-4-iodobenzene, 99%
Thermo Scientific Chemicals
1-Fluoro-4-iodobenzene, 99%
CAS: 352-34-1 | C6H4FI | 222.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
1 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA204500500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Fluoro-4-iodobenzene
CAS352-34-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0