Eicosane, 99%
Eicosane, 99%
Eicosane, 99%
Thermo Scientific Chemicals
Eicosane, 99%
CAS: 112-95-8 | C20H42 | 282.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 g
100 g
Packaging:
Glass bottle
Catalog number FSA204375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEicosane
CAS112-95-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.