N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%
N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%
N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%
N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%
Thermo Scientific Chemicals
N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%
CAS: 536-46-9 | C8H14Cl2N2 | 209.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA204351000
Price (MYR)
2,827.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,827.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethyl-p-phenylenediamine dihydrochloride
CAS536-46-9
CAS Min %98.5
Health Hazard 1Danger
CAS Max %100.0