N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
Thermo Scientific Chemicals
N,N-Dimethyl-1-naphthylamine, 99%
CAS: 86-56-6 | C12H13N | 171.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA204170250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Dimethyl-1-naphthylamine
Name Note99%
CAS86-56-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning