Dicumarol, 99%
Dicumarol, 99%
Dicumarol, 99%
Thermo Scientific Chemicals
Dicumarol, 99%
CAS: 66-76-2 | C19H12O6 | 336.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA204120250
Price (MYR)
915.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
915.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDicumarol
Name Note99%
CAS66-76-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure if swallowed.