4-Nitroquinoline-N-oxide, 98%
4-Nitroquinoline-N-oxide, 98%
4-Nitroquinoline-N-oxide, 98%
4-Nitroquinoline-N-oxide, 98%
Thermo Scientific Chemicals
4-Nitroquinoline-N-oxide, 98%
CAS: 56-57-5 | C9H6N2O3 | 190.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA203792500
Price (MYR)
387.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
387.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Nitroquinoline-N-oxide
CAS56-57-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0