DL-3-Octanol, 97%
DL-3-Octanol, 97%
DL-3-Octanol, 97%
Thermo Scientific Chemicals
DL-3-Octanol, 97%
CAS: 589-98-0 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA203772500
Price (MYR)
1,767.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,767.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-3-Octanol
CAS589-98-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.