Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
CAS: 12612-37-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic Bottle
Catalog number FSA203041000
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 50WX2
Physical FormBeads
CAS12612-37-2
Particle SizeParticle size analysis: on 70 mesh (cumulative): <15%, through 270 mesh: <15%
PackagingPlastic Bottle