Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number FSA202965000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 1X2
Physical FormBeads
CAS9085-42-1
Particle SizeParticle size analysis: on 50 mesh (cumulative): < 15%, through 140 mesh: < 15%
PackagingPlastic Bottle