Dowex™ 1X2, 50-100 mesh, ion-exchange resin
Dowex™ 1X2, 50-100 mesh, ion-exchange resin
Dowex™ 1X2, 50-100 mesh, ion-exchange resin
Dowex™ 1X2, 50-100 mesh, ion-exchange resin
Dowex™ 1X2, 50-100 mesh, ion-exchange resin
CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA202955000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 1X2
Physical FormBeads
CAS9085-42-1
PackagingPlastic bottle
Molecular FormulaC29H34ClN
View more
Ion exchange resin
RUO – Research Use Only