Trichloroacetyl isocyanate, NMR grade
Trichloroacetyl isocyanate, NMR grade
Trichloroacetyl isocyanate, NMR grade
Trichloroacetyl isocyanate, NMR grade
Thermo Scientific Chemicals
Trichloroacetyl isocyanate, NMR grade
CAS: 3019-71-4 | C3Cl3NO2 | 188.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Packaging:
Glass bottle
Catalog number FSA202900500
Price (MYR)
2,885.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,885.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrichloroacetyl isocyanate
CAS3019-71-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Reacts violently with water.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
For NMR
RUO – Research Use Only