Potassium sodium tartrate tetrahydrate, 99+%, for analysis
Potassium sodium tartrate tetrahydrate, 99+%, for analysis
Potassium sodium tartrate tetrahydrate, 99+%, for analysis
Potassium sodium tartrate tetrahydrate, 99+%, for analysis
Thermo Scientific Chemicals
Potassium sodium tartrate tetrahydrate, 99+%, for analysis
CAS: 6381-59-5 | C4H4KNaO6·4H2O | 282.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA202865000
Price (MYR)
520.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
520.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium sodium tartrate tetrahydrate
CAS6381-59-5
Decomposition Information>100°C
ColorWhite
pH5.5 to 8.5 (5% soln. at 20°C)