4',6'-Diamidino-2-phenylindole dihydrochloride, 98%
4',6'-Diamidino-2-phenylindole dihydrochloride, 98%
4',6'-Diamidino-2-phenylindole dihydrochloride, 98%
4',6'-Diamidino-2-phenylindole dihydrochloride, 98%
Thermo Scientific Chemicals
4',6'-Diamidino-2-phenylindole dihydrochloride, 98%
CAS: 28718-90-3 | C16H17Cl2N5 | 350.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Packaging:
Glass bottle
Catalog number FSA202710500
Price (MYR)
2,063.00
EA
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,063.00
EA
Add to cart
Specifications
Chemical Name or Material4′,6′-Diamidino-2-phenylindole dihydrochloride
CAS28718-90-3
CAS Min %97.5
CAS Max %100.0
EINECS Number249-186-7