1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
Thermo Scientific Chemicals
1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
CAS: 64431-96-5 | C11H26N2O6 | 282.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA202641000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,3-Bis[tris(hydroxymethyl)amino]propane
CAS64431-96-5
CAS Min %98.5
CAS Max %100.0
Melting Point164.0°C to 165.0°C