Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized
CAS: 7722-84-1 | H2O2 | 34 g/mol
Have Questions?
Change view
Quantity:
500 mL
1 L
2.5 L
Packaging:
Plastic bottle
Catalog number FSA202465000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point108.0°C
Chemical Name or MaterialHydrogen peroxide
Melting Point-33.0°C
Name NoteFor analysis, Stabilized
CAS7722-84-1