Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Thermo Scientific Chemicals
Tetrakis(triphenylphosphine)palladium(0), 99%
CAS: 14221-01-3 | C72H60P4Pd | 1155.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA202380050
Price (MYR)
1,331.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,331.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetrakis(triphenylphosphine)palladium(0)
CAS14221-01-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.