Ammonium hexafluorophosphate, 99%, pure
Ammonium hexafluorophosphate, 99%, pure
Ammonium hexafluorophosphate, 99%, pure
Ammonium hexafluorophosphate, 99%, pure
Thermo Scientific Chemicals
Ammonium hexafluorophosphate, 99%, pure
CAS: 16941-11-0 | H4F6NP | 163 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Plastic bottle
Catalog number FSA202321000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hexafluorophosphate
Melting Point198°C
Name Note99.5%
CAS16941-11-0
ColorWhite to Beige