Amberlite™ XAD-7 HP
Amberlite™ XAD-7 HP
Amberlite™ XAD-7 HP
Amberlite™ XAD-7 HP
Thermo Scientific Chemicals
Amberlite™ XAD-7 HP
CAS: 37380-43-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA202245000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ XAD-7 HP
CAS37380-43-1
PackagingPlastic bottle
MDL NumberMFCD00132705
Quantity500 g
View more
Ion exchange resin
RUO – Research Use Only