Amberlite™ XAD-4
Amberlite™ XAD-4
Amberlite™ XAD-4
Thermo Scientific Chemicals
Amberlite™ XAD-4
CAS: 9003-69-4 | C10H10
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA202235000
Price (MYR)
917.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
917.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmberlite™ XAD-4
Physical FormCrystalline Powder or Crystals
CAS9003-69-4
Particle Size2.0 (uniformity coefficient) Max. 0.490 to 0.690mm (harmonic mean size) 5.0% Max. (<0.300mm) 5.0% Max. (>1.180 mm)
PackagingPlastic bottle
View more
Ion exchange resin
RUO – Research Use Only