Hydrazine sulfate, 99+%
Hydrazine sulfate, 99+%
Hydrazine sulfate, 99+%
Hydrazine sulfate, 99+%
Thermo Scientific Chemicals
Hydrazine sulfate, 99+%
CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA201865000
Price (MYR)
933.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
933.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydrazine sulfate
Name Notep.a.
CAS10034-93-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause cancer.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
View more
Reduction reagent
RUO – Research Use Only