Ammonium dihydrogen phosphate, 99+%, for analysis
Ammonium dihydrogen phosphate, 99+%, for analysis
Ammonium dihydrogen phosphate, 99+%, for analysis
Ammonium dihydrogen phosphate, 99+%, for analysis
Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, 99+%, for analysis
CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA201832500
Price (MYR)
429.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
429.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium dihydrogen phosphate
Melting Point190.0°C
CAS7722-76-1
ColorWhite
pH4 to 4.5 (5% soln. at 20°C)