Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Thermo Scientific Chemicals
Ammonium phosphate, dibasic, 99+%, for analysis
CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
Packaging:
Plastic bottle
Catalog number FSA201825000
Price (MYR)
608.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
608.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium phosphate, dibasic
Name Notep.a.
CAS7783-28-0
ColorWhite
Health Hazard 1Warning
View more
Suitable for battery materials development.
RUO – Research Use Only