Lead(II) acetate trihydrate, 99+%, for analysis
Lead(II) acetate trihydrate, 99+%, for analysis
Lead(II) acetate trihydrate, 99+%, for analysis
Lead(II) acetate trihydrate, 99+%, for analysis
Thermo Scientific Chemicals
Lead(II) acetate trihydrate, 99+%, for analysis
CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Plastic bottle
Catalog number FSA201755000
Price (MYR)
532.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
532.00
EA
Add to cart
Specifications
Chemical Name or MaterialLead(II) acetate trihydrate
Melting Point75.0°C
Name Notep.a.
CAS6080-56-4