Antimony trichloride, 99.5%
Antimony trichloride, 99.5%
Antimony trichloride, 99.5%
Antimony trichloride, 99.5%
Thermo Scientific Chemicals
Antimony trichloride, 99.5%
CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA201685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point223°C
Chemical Name or MaterialAntimony trichloride
Melting Point73°C
CAS10025-91-9