Ammonium cerium(IV) nitrate, 99%, for analysis
Ammonium cerium(IV) nitrate, 99%, for analysis
Ammonium cerium(IV) nitrate, 99%, for analysis
Ammonium cerium(IV) nitrate, 99%, for analysis
Thermo Scientific Chemicals
Ammonium cerium(IV) nitrate, 99%, for analysis
CAS: 16774-21-3 | H8CeN8O18 | 548.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA201442500
Price (MYR)
871.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
871.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium cerium(IV) nitrate
Melting Point107.0°C to 108.0°C
Name Notep.a.
CAS16774-21-3
ColorColorless to Yellow
View more
Oxidation reagent for analysis
RUO – Research Use Only