1-Methyladenine, 95%, Thermo Scientific Chemicals
1-Methyladenine, 95%, Thermo Scientific Chemicals
1-Methyladenine, 95%, Thermo Scientific Chemicals
1-Methyladenine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyladenine, 95%, Thermo Scientific Chemicals
CAS: 5142-22-3 | C6H7N5 | 149.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
250 mg
Packaging:
Glass bottle
Catalog number FSA201311000
Price (MYR)
1,699.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,699.00
EA
Add to cart
Specifications
Chemical Name or Material1-Methyladenine
CAS5142-22-3
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0