Sulfur, 99.5%, sublimed
Sulfur, 99.5%, sublimed
Sulfur, 99.5%, sublimed
Sulfur, 99.5%, sublimed
Thermo Scientific Chemicals
Sulfur, 99.5%, sublimed
CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA201250250
Price (MYR)
298.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
298.00
BOT
Add to cart
Specifications
Boiling Point445.0°C
Chemical Name or MaterialSulfur
Melting Point114.0°C
Name Note99.5%
CAS7704-34-9