Silicon(IV) chloride, 99.8+%
Silicon(IV) chloride, 99.8+%
Silicon(IV) chloride, 99.8+%
Silicon(IV) chloride, 99.8+%
Thermo Scientific Chemicals
Silicon(IV) chloride, 99.8+%
CAS: 10026-04-7 | Cl4Si | 169.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA201165000
Price (MYR)
659.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
659.00
BOT
Add to cart
Specifications
Boiling Point58.0°C
Chemical Name or MaterialSilicon(IV) chloride
Melting Point-70.0°C
Name Note99.8+%
CAS10026-04-7