Hypophosphorous acid, 50 wt.% solution in wate
Hypophosphorous acid, 50 wt.% solution in wate
Hypophosphorous acid, 50 wt.% solution in wate
Hypophosphorous acid, 50 wt.% solution in wate
Thermo Scientific Chemicals
Hypophosphorous acid, 50 wt.% solution in wate
CAS: 6303-21-5 | H3O2P | 66.00 g/mol
Have Questions?
Catalog NumberQuantity
FSA2010000101 L
Catalog number FSA201000010
Price (MYR)
782.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
782.00
EA
Add to cart
Specifications
Chemical Name or MaterialHypophosphorous acid
CAS6303-21-5
Melting Point-25.0°C
Boiling Point108.0°C (1013.0 hPa)
Density1.27 g/mL