Sulfur, 99.999%, (trace metal basis)
Sulfur, 99.999%, (trace metal basis)
Sulfur, 99.999%, (trace metal basis)
Sulfur, 99.999%, (trace metal basis)
Thermo Scientific Chemicals
Sulfur, 99.999%, (trace metal basis)
CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA199930500
Price (MYR)
832.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
832.00
BOT
Add to cart
Specifications
Boiling Point445.0°C
Chemical Name or MaterialSulfur
Melting Point114.0°C
Name Note99.999%
CAS7704-34-9
View more
For ultra trace analysis
RUO – Research Use Only