Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh
Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh
Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh
Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh
Thermo Scientific Chemicals
Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh
CAS: 13759-92-7 | Cl3Eu·6H2O | 366.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA199600050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEuropium(III) chloride hexahydrate
Melting Point850.0°C
Name Note99.9%
CAS13759-92-7
ColorWhite
View more
for ultra trace analysis
RUO – Research Use Only