Vanadyl(IV) acetylacetonate, 99%
Vanadyl(IV) acetylacetonate, 99%
Vanadyl(IV) acetylacetonate, 99%
Vanadyl(IV) acetylacetonate, 99%
Thermo Scientific Chemicals
Vanadyl(IV) acetylacetonate, 99%
CAS: 3153-26-2 | C10H14O5V | 265.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA199590500
Price (MYR)
569.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
569.00
EA
Add to cart
Specifications
Chemical Name or MaterialVanadyl(IV) acetylacetonate
Name Note99%
CAS3153-26-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement