Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals
Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
CAS: 16853-85-3 | AlH4Li | 37.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA199511000
Price (MYR)
683.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
683.00
EA
Add to cart
Chemical Identifiers
CAS16853-85-3, 60-29-7
Specifications
Chemical Name or MaterialLithium Aluminum hydride
ColorColorless
Name Note4.0M Solution in Diethyl Ether
CAS16853-85-3
Health Hazard 1Danger
View more
Reduction reagent
RUO – Research Use Only