Sulfamethazine, 99%, Thermo Scientific Chemicals
Sulfamethazine, 99%, Thermo Scientific Chemicals
Sulfamethazine, 99%, Thermo Scientific Chemicals
Sulfamethazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfamethazine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA199301000
Price (MYR)
509.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
509.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSulfamethazine
Name Note99%
CAS57-68-1
Health Hazard 1LIGHT SENSITIVE
Melting Point197.0°C to 200.0°C