Cerium(IV) sulfate tetrahydrate, 98+%
Cerium(IV) sulfate tetrahydrate, 98+%
Cerium(IV) sulfate tetrahydrate, 98+%
Cerium(IV) sulfate tetrahydrate, 98+%
Thermo Scientific Chemicals
Cerium(IV) sulfate tetrahydrate, 98+%
CAS: 10294-42-5 | CeO8S2·4H2O | 404.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA199135000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(IV) sulfate tetrahydrate
Name Note99+%
CAS10294-42-5
ColorYellow
Health Hazard 1GHS Signal Word: Danger