Cerium(IV) oxide, 99.9%, (trace metal basis)
Cerium(IV) oxide, 99.9%, (trace metal basis)
Cerium(IV) oxide, 99.9%, (trace metal basis)
Cerium(IV) oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Cerium(IV) oxide, 99.9%, (trace metal basis)
CAS: 1306-38-3 | CeO2 | 172.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA199125000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(IV) oxide
Melting Point2600.0°C
Name Note99.9%
CAS1306-38-3
ColorWhite to Yellow
View more
For ultra trace analysis
RUO – Research Use Only