Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Thermo Scientific Chemicals
Hydroxylamine sulfate, 99%
CAS: 10039-54-0 | H6N2O2·H2SO4 | 164.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA198531000
Price (MYR)
251.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
251.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydroxylamine sulfate
Melting Point170.0°C
CAS10039-54-0
ColorWhite
Health Hazard 1GHS Signal Word: Warning