2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
CAS: 2033-24-1 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA198451000
Price (MYR)
734.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
734.00
BOT
Add to cart
Specifications
Chemical Name or Material2,2-Dimethyl-1,3-dioxane-4,6-dione
Name Note98%
CAS2033-24-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning