Bromoacetyl chloride, 95%
Bromoacetyl chloride, 95%
Bromoacetyl chloride, 95%
Bromoacetyl chloride, 95%
Thermo Scientific Chemicals
Bromoacetyl chloride, 95%
CAS: 22118-09-8 | C2H2BrClO | 157.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA197990250
Price (MYR)
533.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
533.00
EA
Add to cart
Specifications
Chemical Name or MaterialBromoacetyl chloride
CAS22118-09-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat