4-Fluorobenzyl bromide, 97%
4-Fluorobenzyl bromide, 97%
4-Fluorobenzyl bromide, 97%
4-Fluorobenzyl bromide, 97%
Thermo Scientific Chemicals
4-Fluorobenzyl bromide, 97%
CAS: 459-46-1 | C7H6BrF | 189.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA197970250
Price (MYR)
540.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
540.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Fluorobenzyl bromide
Name Note97%
CAS459-46-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger