2-Fluorobenzyl bromide, 98%
2-Fluorobenzyl bromide, 98%
2-Fluorobenzyl bromide, 98%
2-Fluorobenzyl bromide, 98%
Thermo Scientific Chemicals
2-Fluorobenzyl bromide, 98%
CAS: 446-48-0 | C7H6BrF | 189.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA197960250
Price (MYR)
767.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
767.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Fluorobenzyl bromide
Name Note98%
CAS446-48-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger