Urea, 98%, extra pure, pearls
Urea, 98%, extra pure, pearls
Urea, 98%, extra pure, pearls
Urea, 98%, extra pure, pearls
Thermo Scientific Chemicals
Urea, 98%, extra pure, pearls
CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number FSA197460050
Price (MYR)
747.00
BOT
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
747.00
BOT
Add to cart
Specifications
Chemical Name or MaterialUrea
Name Note98%
CAS57-13-6
Health Hazard 2GHS H Statement:
Causes skin irritation.
Health Hazard 3GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.