Copper(I) oxide, 97%
Copper(I) oxide, 97%
Copper(I) oxide, 97%
Copper(I) oxide, 97%
Thermo Scientific Chemicals
Copper(I) oxide, 97%
CAS: 1317-39-1 | Cu2O | 143.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Glass bottle
Catalog number FSA197425000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point1800.0°C
Chemical Name or MaterialCopper(I) oxide
Melting Point1232.0°C
Name Note97%
CAS1317-39-1