S-Methyl methanethiolsulfonate, 97%
S-Methyl methanethiolsulfonate, 97%
S-Methyl methanethiolsulfonate, 97%
S-Methyl methanethiolsulfonate, 97%
Thermo Scientific Chemicals
S-Methyl methanethiolsulfonate, 97%
CAS: 2949-92-0 | C2H6O2S2 | 126.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA197390050
Price (MYR)
545.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
545.00
BOT
Add to cart
Specifications
Chemical Name or MaterialS-Methyl methanethiolsulfonate
CAS2949-92-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at