n-Hexane, 95+%, extra pure
n-Hexane, 95+%, extra pure
n-Hexane, 95+%, extra pure
n-Hexane, 95+%, extra pure
Thermo Scientific Chemicals
n-Hexane, 95+%, extra pure
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 L
Packaging:
View all
Metal Drum
Catalog number FSA197360250
Price (MYR)
3,813.00
BOT
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
3,813.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
Name Note95%
CAS110-54-3
CAS Min %95.0