Molecular sieves 5A, 4 to 8 mesh
Molecular sieves 5A, 4 to 8 mesh
Molecular sieves 5A, 4 to 8 mesh
Molecular sieves 5A, 4 to 8 mesh
Thermo Scientific Chemicals
Molecular sieves 5A, 4 to 8 mesh
CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA197285000
Price (MYR)
342.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
342.00
BOT
Add to cart
Specifications
Bulk Density660 to 780g/L
Chemical Name or MaterialMolecular sieves 5A
ColorBeige
PackagingGlass bottle
Particle Size4 to 8 mesh
View more
Drying agent
RUO – Research Use Only