Molecular sieves 4A, 4 to 8 mesh
Molecular sieves 4A, 4 to 8 mesh
Molecular sieves 4A, 4 to 8 mesh
Molecular sieves 4A, 4 to 8 mesh
Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA197265000
Price (MYR)
490.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
490.00
BOT
Add to cart
Specifications
Bulk Density660 to 760g/L
Chemical Name or MaterialMolecular sieves 4A
ColorColorless to Yellow
PackagingGlass bottle
Residual Water1.5% max.
View more
Drying agent
RUO – Research Use Only