Molecular sieves 3A, 8 to 12 mesh
Molecular sieves 3A, 8 to 12 mesh
Molecular sieves 3A, 8 to 12 mesh
Molecular sieves 3A, 8 to 12 mesh
Thermo Scientific Chemicals
Molecular sieves 3A, 8 to 12 mesh
CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 kg
Catalog number FSA197255000
Price (MYR)
641.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
641.00
BOT
Add to cart
Specifications
Bulk Density600g/L min.
Chemical Name or MaterialMolecular sieves 3A
ColorBeige to Brown
PackagingGlass bottle
Residual Water1.5% max.
View more
Drying agent
RUO – Research Use Only