Molecular sieves 3A, 4 to 8 mesh
Molecular sieves 3A, 4 to 8 mesh
Molecular sieves 3A, 4 to 8 mesh
Molecular sieves 3A, 4 to 8 mesh
Thermo Scientific Chemicals
Molecular sieves 3A, 4 to 8 mesh
CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA197245000
Price (MYR)
546.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
546.00
BOT
Add to cart
Specifications
Bulk Density700 to 800g/L
Chemical Name or MaterialMolecular sieves 3A
ColorBeige
PackagingGlass bottle
Residual Water1.5% max. (550°C, 2 hrs.)
View more
Drying agent
RUO – Research Use Only